Platinum rhodium alloy (94:6 w%), wire for ISA type B standard grade thermocouple, Ø 0.33 mm (0.013 in)
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
43025.G2
43025.BW
43025.G2EA
1964
EUR
43025.G2
43025.BW
Platinum rhodium alloy (94:6 w%), wire for ISA type B standard grade thermocouple, Ø 0.33 mm (0.013 in)
Platinum rhodium alloy (94:6 w%)
Molecular Formula: Pt:Rh; 94:6 wt%
Physical Form: ≈0.88g/50cm
Odour: Odourless
MDL No.: MFCD00198186