Platinum rhodium alloy (95.2:4.8 w%), foil, Thickness: 0.05 mm (0.002 in)
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
42686.FF
42686.FI
42686.FFEA
209
EUR
42686.FF
42686.FI
Platinum rhodium alloy (95.2:4.8 w%), foil, Thickness: 0.05 mm (0.002 in)
Platinum rhodium alloy (95.2:4.8 w%)
Molecular Formula: Pt:Ru; 95.2:4.8 wt%
Physical Form: ≈0.66g/25x25mm
Odour: Odourless
MDL No.: MFCD00798552