Aluminium standard solution, 1000 mg/L Al in 0,5 M nitric acid (from Al(NO₃)₃), standard for AAS, Certipur®, Supelco®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1.19770.0100 1.19770.0500
1.19770.0100EA 40.6 EUR
1.19770.0100 1.19770.0500
Aluminium standard solution, 1000 mg/L Al in 0,5 M nitric acid (from Al(NO₃)₃), standard for AAS, Certipur®, Supelco®
Standaarden Kationische standaarden Aluminium standaarden
Certificaten: Traceable to SRM from NIST.