VWR®, Ammonium standard solution, 1000 mg/L NH₄ in water, ARISTAR® standard for Ion Chromatography
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
458192MEA 166 EUR
458192M 458194G
VWR®, Ammonium standard solution, 1000 mg/L NH₄ in water, ARISTAR® standard for Ion Chromatography
Standaarden Kationische standaarden Ammonium standaarden
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.