Barium standard solution, 1000 mg/L Ba in 0,5 M nitric acid (from Ba(NO₃)₂) Certipur® standard for AAS, Supelco®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1.19774.0100 1.19774.0500
1.19774.0100EA 36.8 EUR
1.19774.0100 1.19774.0500
Barium standard solution, 1000 mg/L Ba in 0,5 M nitric acid (from Ba(NO₃)₂) Certipur® standard for AAS, Supelco®
Standaarden Kationische standaarden Barium standaarden
Certificaten: Traceable to SRM from NIST.