VWR® ARISTAR®, Chromium Standard Solutions, Standards for ICP
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
455242QEA 94.8 EUR
455242Q 455254U 455252S 455244S 85762.180 85762.270
VWR® ARISTAR®, Chromium Standard Solutions, Standards for ICP
Standaarden Kationische standaarden Chroom standaarden
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.