VWR® ARISTAR®, Cesium Standard Solution, 1000 mg/L Cs in dil. Nitric Acid (from CsNO₃), Standard for ICP
Leverancier: VWR Chemicals
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
455204KEA
240
EUR
455204K
455202Y
VWR® ARISTAR®, Cesium Standard Solution, 1000 mg/L Cs in dil. Nitric Acid (from CsNO₃), Standard for ICP
Standaarden
Kationische standaarden
Cesium standaarden
CsNO₃ in HNO₃ 2 to 5%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.