VWR®, Dysprosium standard solution, 1000 mg/L Dy in dil. nitric acid (from Dy₂O₃) ARISTAR® standard for ICP
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
455302LEA 57.66 EUR
455302L 455304N
VWR®, Dysprosium standard solution, 1000 mg/L Dy in dil. nitric acid (from Dy₂O₃) ARISTAR® standard for ICP
Standaarden Kationische standaarden Dysprosium standaarden
Dy₂O₃ in 2 to 5% HNO₃.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.