VWR® ARISTAR®, Magnesium Standard Solution, 10000 mg/L Mg in dil. Nitric Acid (from MgO), Standard for ICP
Leverancier: VWR Chemicals
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
455632DEA
281
EUR
455632D
455634F
VWR® ARISTAR®, Magnesium Standard Solution, 10000 mg/L Mg in dil. Nitric Acid (from MgO), Standard for ICP
Standaarden
Kationische standaarden
Magnesium standaarden
MgO in HNO₃ 2 to 5%.
Certificaten: Traceable to SRM from NIST, manufactured and tested in ISO 17034 / ISO 17025 accredited facilities.
Leveringsinformatie: Supplied with certificate of analysis.