ARISTAR®VWR® , Manganese Standard Solution, 1000 mg/L Mn in dil. Nitric Acid (from Mn(OAc)₂), Standard for ICP
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
455642FEA 160 EUR
455642F 455644H
ARISTAR®VWR® , Manganese Standard Solution, 1000 mg/L Mn in dil. Nitric Acid (from Mn(OAc)₂), Standard for ICP
Standaarden Kationische standaarden Mangaan standaarden
Mn(C₂H₃O₂)₂ in HNO₃ 2 to 5%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.