VWR® ARISTAR®, Manganese Standard Solution, 10000 mg/L Mn in dil. Nitric Acid (from Mn(OAc)₂), Standard for ICP
Leverancier: VWR Chemicals
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
455652HEA
316
EUR
455652H
455654J
VWR® ARISTAR®, Manganese Standard Solution, 10000 mg/L Mn in dil. Nitric Acid (from Mn(OAc)₂), Standard for ICP
Standaarden
Kationische standaarden
Mangaan standaarden
Mn(C₂H₃O₂)₂ in HNO₃ 2 to 5%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.