VWR® AVS TITRINORM, Silver Standard Solution, 1000 mg/L Ag in dil. Nitric Acid, Standard for AAS
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
86659.260EA 36.3 EUR
86659.260 86659.180
VWR® AVS TITRINORM, Silver Standard Solution, 1000 mg/L Ag in dil. Nitric Acid, Standard for AAS
Standaarden Kationische standaarden Zilver standaarden
  • Traceable to SRM from NIST
  • Manufactured and tested in ISO Guide 34/ISO 17025 accredited facilities
  • Supplied with Certificate of Analysis, real value ±0,3%