Polyethylene glycol 300 for synthesis, Sigma-Aldrich®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M-Clarity™ quality level = MQ200
8.07484.1000 8.07484.5000
8.07484.1000EA 53.9 EUR
8.07484.1000 8.07484.5000
Polyethylene glycol 300 for synthesis, Sigma-Aldrich®
Polyethylene glycol 300
Formule: H(OCH₂CH₂)nOH
Smeltpunt: –15…–10 °C
Dichtheid: 1,13 g/cm³ (20 °C)
Vlampunt: 220 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00081839
CAS nummer: 25322-68-3
EINECS: 500-038-2
Order Now
Specificaties
Environmentally Preferable