Polyethylene glycol 400, GPR RECTAPUR®
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: PEG 400
26602.290EA 31.8 EUR
26602.290 26602.460 26602.320
Polyethylene glycol 400, GPR RECTAPUR®
Polyethylene glycol 400
Formule: H(OCH₂CH₂)nOH
Kookpunt: >250 °C (1013 hPa)
Smeltpunt: 4…8 °C
Dichtheid: 1,13 g/cm³ (20 °C)
Vlampunt: 245 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00081839
CAS nummer: 25322-68-3
EINECS: 500-038-2
Order Now
Specification Test Results
Identification Passes test
Acid value (mg KOH/g) Max. 0.2
Density (20/4) 1.120 - 1.130
Hydroxyl value 200 - 300