Polyethylene glycol 40,000
Leverancier: Serva
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
33139.01
33139.02
SERA33139.01EA
34
EUR
SERA33139.01
SERA33139.02
Polyethylene glycol 40,000
Polyethylene glycol 40,000
Formule:
H(OCH₂CH₂)nOH MW: 40000 g/mol Bewaar temperatuur: Kamertemperatuur |
CAS nummer:
25322-68-3 EINECS: 500-038-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...