Polyethylene glycol 6,000, EMPROVE® ESSENTIAL Ph. Eur. suitable for use as excipient, SAFC®
Leverancier: MERCK PRODUCTION CHEMICALS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: PEG 6,000, PEG 6000
8.17007.5000 8.17007.9050
8.17007.5000EA 413 EUR
8.17007.5000 8.17007.9050
Polyethylene glycol 6,000, EMPROVE® ESSENTIAL Ph. Eur. suitable for use as excipient, SAFC®
Polyethylene glycol 6,000
Formule: H(OCH₂CH₂)nOH
Kookpunt: ∼250 °C (1013 hPa)
Smeltpunt: 55…62 °C
Dichtheid: 1,13 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD01779614
CAS nummer: 25322-68-3
EINECS: 500-038-2