Polyethylene glycol 8,000
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
418050010 418050030 418050100
ACRO418050010EA 80.2 EUR
ACRO418050010 ACRO418050030 ACRO418050100
Polyethylene glycol 8,000
Polyethylene glycol 8,000
Formule: H(OCH₂CH₂)nOH
Kookpunt: 250 °C (1013 hPa)
Smeltpunt: 55…60 °C
Dichtheid: 1,125 g/cm³ (20 °C)
Vlampunt: 270 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD01779615
CAS nummer: 25322-68-3
EINECS: 500-038-2
Merck Index: 12,07729
Order Now
Specification Test Results
Appearance White Flakes
Infrared spectrum Conforms
Water ≤0.5 % (K.F.)
Hydroxyl value 12 to 16 mg KOH/g