di-Sodium L(+)-tartrate dihydrate 99.0-101.0% ACS
Leverancier: Thermo Fisher ScientificTotal Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen:
Sodium L-(+)-tartrate dihydrate, L(+)-Tartaric acid disodium salt dihydrate, (R,R)-(+)-Tartaric acid disodium salt dihydrate, (R,R)-(+)-Sodium tartrate dibasic dihydrate, Disodium L-(+)-Tartrate Dihydrate, (R,R)-(+)-Disodium tartrate dihydrate, L(+)-Tartaric acid, disodium salt, dihydrate, Sodium L-tartrate dibasic dihydrate, L(+)-Sodium tartrate dibasic dihydrate, Sodium Tartrate, Dihydrate, (+)-Disodium L-tartrate dihydrate, (+)-Disodium-L-tartrat dihydrate
Purity: 99.0-101.0%
CAS No.: 6106-24-7
Molecular Formula: C4H4Na2O6.2H2O
Formula Weight: 230.08 (194.06anhy)
Physical Form: Granular
Density: 1.82
Melting Point: ca 150° -2H2O
Solubility: Soluble in water
Beilstein Reference: 6121732
Merck Reference: 14,8684
MDL No.: MFCD00150035
Formule:
NaOOC(CHOH)₂COONa·2H₂O MW: 230.08 g/mol Smeltpunt: ∼150 °C (-2H₂O) Dichtheid: 1.82 Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00150035 CAS nummer: 6106-24-7 EINECS: 212-773-3 Merck Index: 14,8684 |