Tannine
Leverancier: Roth Carl
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen:
Gallotannin, Tannin
4239.1
4239.2
ROTH4239.1EA
0
EUR
ROTH4239.1
ROTH4239.2
Tannine
Tannine
Formule:
C₇₆H₅₂O₄₆ MW: 1701.2 g/mol Smeltpunt: 218…219 °C Dichtheid: Vlampunt: 199 |
MDL Nummer:
MFCD00066397 CAS nummer: 1401-55-4 EINECS: 215-753-2 Merck Index: 12,09221 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...