Polyvinylpyrrolidone, M.W. ∼ 1.300.000 (K85-95)
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen:
Povidone, Polyvinylpyrrolidon, Polyvidone, PVP
43728.A1
43728.30
43728.18
43728.A1EA
235
EUR
43728.A1
43728.30
43728.18
Polyvinylpyrrolidone, M.W. ∼ 1.300.000 (K85-95)
Polyvinylpyrrolidone
CAS No.: 9003-39-8
Molecular Formula: (C6H9NO)n
Physical Form: Powder
Sensitivity: Hygroscopic
Odour: Odourless
Merck Reference: 14,7697
MDL No.: MFCD00149016
Formule:
(C₆H₉NO)n Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD01076626 CAS nummer: 9003-39-8 Merck Index: 14,7697 |