Polyvinylpyrrolidone, M.W. ∼ 360,000 (K90)

Leverancier: Serva
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: Povidone, Polyvinylpyrrolidon, Polyvidone, PVP

39609.01 39609.02
SERA39609.01EA 0 EUR
SERA39609.01 SERA39609.02
Polyvinylpyrrolidone, M.W. ∼ 360,000 (K90)
Polyvinylpyrrolidone
Formule: (C₆H₉NO)n
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD01076626
CAS nummer: 9003-39-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR