Methyleenchloride 99,7 Stabilised Spectrophotometry
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
Methylene chloride
Stabilisatie:
Amylen
32440.K2
32440.M1
32440.K2EA
62.2
EUR
32440.K2
32440.M1
Methyleenchloride 99,7 Stabilised Spectrophotometry
Methyleenchloride
Purity: 99.7+%
CAS No.: 75-09-2
Molecular Formula: CH2Cl2
Formula Weight: 84.93
Physical Form: Liquid
Density: 1.325
Refractive Index: 1.4244
Odour: Sweet
Melting Point: -95°
Boiling Point: 39-40°
Beilstein Reference: 1730800
Merck Reference: 14,6063
UN No.: 1593
MDL No.: MFCD00000881
Formule:
CH₂Cl₂ MW: 84.93 g/mol Kookpunt: 39…40 °C Smeltpunt: –95 °C Dichtheid: 1.325 Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00000881 CAS nummer: 75-09-2 EINECS: 200-838-9 UN: 1593 ADR: 6.1 III Merck Index: 14,6063 |