(±)-Linalool for synthesis, Sigma-Aldrich®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
M-Clarity™ quality level = MQ200
8.18627.0005 8.18627.0250
8.18627.0005EA 33.2 EUR
8.18627.0005 8.18627.0250
(±)-Linalool for synthesis, Sigma-Aldrich®
(±)-Linalool
Formule: C₁₀H₁₈O
Kookpunt: 199 °C (1013 hPa)
Dichtheid: 0,87 g/cm³ (20 °C)
Vlampunt: 75 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008906
CAS nummer: 78-70-6
EINECS: 201-134-4
Merck Index: 13,05517
Order Now
Specification Test Results
Solubility 1.45 g/l (20 °C)
Molar Mass 154.25 g/mol
Boiling Point 198 - 200 °C
Vapor Pressure 0.10 hPa (20 °C)
Flash Point 79 °C
Refractive Index 1.462 (20 °C, 589 nm)
Explosion Limit 0.9 - 5.2 %(V)
Density 0.86 g/cm3 (20 °C)
pH 4.5 (1.45 g/l, H2O, 25 °C)
Ignition Point 235 °C