(±)-2-Bromobutyric acid 99%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: 2-Bromobutanoic acid, (±)-2-Bromobutanoic acid
164510010 164512500 164510050
ACRO164510010EA 338 EUR
ACRO164510010 ACRO164512500 ACRO164510050
(±)-2-Bromobutyric acid 99%
(±)-2-Bromobutyric acid
Formule: H₃CCH₂BrCHCOOH
MW: 167 g/mol
Kookpunt: 214
Smeltpunt: –4 °C
Dichtheid: 1.56 g/cm³
Vlampunt: >112 °C
MDL Nummer: MFCD00004216
CAS nummer: 80-58-0
EINECS: 201-294-5
UN: 2922
Merck Index: 13,01397
Order Now
Specification Test Results
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4715 to 1.4755 (20°C, 589 nm)
GC ≥98.5 %