(±)-2-Bromopropionic acid 99%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
107350010 107352500
ACRO107350010EA 167 EUR
ACRO107350010 ACRO107352500
(±)-2-Bromopropionic acid 99%
(±)-2-Bromopropionic acid
Formule: H₃CBrCHCOOH
Kookpunt: 203 °C (760 mmHg)
Smeltpunt: ∼25 °C
Dichtheid: 1,65 g/cm³
Vlampunt: 100 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004211
CAS nummer: 598-72-1
EINECS: 209-947-6
UN: 3261
ADR: 8,II
Order Now
Specification Test Results
Appearance Clear colorless to yellow Liquid after melting
Infrared spectrum Conforms
Refractive index 1.4740 to 1.4760 (20°C, 589 nm)
HPLC ≥98.5 %
Stabilizer 90 to 150 ppm MEHQ