(±)-2-Chloorpropionzuur

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

SB01780.1 SB01780.10
MAYBSB01780.1EA 0 EUR
MAYBSB01780.1 MAYBSB01780.10
(±)-2-Chloorpropionzuur
(±)-2-Chloorpropionzuur
Formule: C₃H₅ClO₂
Kookpunt: 180 °C (1013 hPa)
Smeltpunt: –12 °C
Dichtheid: 1,27 g/cm³ (20 °C)
MDL Nummer: MFCD00004224
CAS nummer: 598-78-7
EINECS: 209-952-3
UN: 2511
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR