(±)-2-Chloorpropionzuur for synthesis, Sigma-Aldrich®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
M-Clarity™ quality level = MQ200
8.02810.0100 8.02810.0500 8.02810.9025
8.02810.0100EA 37.6 EUR
8.02810.0100 8.02810.0500 8.02810.9025
(±)-2-Chloorpropionzuur for synthesis, Sigma-Aldrich®
(±)-2-Chloorpropionzuur
Formule: C₃H₅ClO₂
Kookpunt: 180 °C (1013 hPa)
Smeltpunt: –12 °C
Dichtheid: 1,27 g/cm³ (20 °C)
MDL Nummer: MFCD00004224
CAS nummer: 598-78-7
EINECS: 209-952-3
UN: 2511
ADR: 8,III
Order Now
Specification Test Results
Solubility (20 °C) soluble
Melting Point -12 °C
Molar Mass 108.52 g/mol
Boiling Point 185 - 186 °C (1013 hPa)
Vapor Pressure 4 hPa (60 °C)
Flash Point 107 °C
Refractive Index 1.4345 (20 °C, 589 nm)
Density 1.27 g/cm3 (20 °C)
pH  - 1 (200 g/l, H2O, 20 °C)
Ignition Point 550 °C