DL-Lithium lactate

Leverancier: Serva
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: DL-Lactic acid lithium salt, (±)-2-Hydroxypropanoic acid lithium salt, Lithium 2-hydroxypropanoate, (±)-2-Hydroxypropionic acid lithium salt, (±)-Lactic acid lithium salt

29742.01 29742.02
SERA29742.01EA 0 EUR
SERA29742.01 SERA29742.02
DL-Lithium lactate
DL-Lithium lactate
Formule: H₃CCH(OH)COOLi
Smeltpunt: 300 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00065399
CAS nummer: 867-55-0
EINECS: 212-761-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR