(±)-2-Aminoheptane ≥98.0% for synthesis, Sigma-Aldrich®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
M-Clarity™ quality level = MQ200
8.41027.0025 8.41027.0100 8.41027.0010
8.41027.0025EA 55.2 EUR
8.41027.0025 8.41027.0100 8.41027.0010
(±)-2-Aminoheptane ≥98.0% for synthesis, Sigma-Aldrich®
(±)-2-Aminoheptane
Formule: C₇H₁₇N
Kookpunt: 143 °C (1013 hPa)
Dichtheid: 0,768 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008101
CAS nummer: 123-82-0
EINECS: 204-655-5
UN: 2733
ADR: 3 (8),III
Merck Index: 12,09934
Order Now
Specification Test Results
CAS 123-82-0
Synonyms 2-Heptylamine, 1-Methylhexylamine
Hill Formula C₇H₁₇N
Molar Mass 115.22 g/mol
Boiling point 142 - 144 °C
Density 0.768 g/cm³ (20 °C)
Flash point 54 °C
pH value (10 g/l, H₂O, 20 °C)
Assay (GC, area%) ≥ 98.0 % (a/a)
Identity (IR) passes test