3-Joodpropeen 97% stabilised
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
218540250
218541000
ACRO218540250EA
84
EUR
ACRO218540250
ACRO218541000
3-Joodpropeen 97% stabilised
3-Joodpropeen
Formule:
C₃H₅I Kookpunt: 102…103 °C (760 mmHg) Smeltpunt: –99 °C Dichtheid: 1,83 g/cm³ Vlampunt: 18 °C Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD00001094 CAS nummer: 556-56-9 EINECS: 209-130-4 UN: 1723 ADR: 3,II Merck Index: 13,00291 |
Specification Test Results
Appearance | Clear yellow to orange to red to brown Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5515 to 1.5565 (20°C, 589 nm) |
GC | ≥96.0 % |
Stabilizer | Copper |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...