2-Fenylpropeen 99% stabilised
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
127710010
127711000
127710025
ACRO127710010EA
56.4
EUR
ACRO127710010
ACRO127711000
ACRO127710025
2-Fenylpropeen 99% stabilised
2-Fenylpropeen
Formule:
C₉H₁₀ Kookpunt: 165…169 °C (760 mmHg) Smeltpunt: –23 °C Dichtheid: 0,909 g/cm³ Vlampunt: 45 °C Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD00008859 CAS nummer: 98-83-9 EINECS: 202-705-0 UN: 2303 ADR: 3,III |
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5370 to 1.5390 (20°C, 589 nm) |
GC | ≥98.5 % |
Stabilizer | 15 ppm tert.-butylcatechol |