(±)-1-Ethoxy-2-propanol ≥90% remainder 2-ethoxy-1-propanol
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
1-Ethoxy-2-propanol, (±)-Monopropylene glycol ethyl ether
43306.AP
43306.AY
43306.APEA
106
EUR
43306.AP
43306.AY
(±)-1-Ethoxy-2-propanol ≥90% remainder 2-ethoxy-1-propanol
(±)-1-Ethoxy-2-propanol
Purity: 90+%
CAS No.: 1569-02-4
Molecular Formula: C5H12O2
Formula Weight: 104.15
Flash Point: 42°(108°F)
Physical Form: Liquid
Density: 0.896
Refractive Index: 1.4065
Appearance: Colourless
Melting Point: -100°
Boiling Point: 132°
UN No.: 1987
MDL No.: MFCD00067050
Formule:
C₅H₁₂O₂ Kookpunt: 132 °C Smeltpunt: –100 °C Dichtheid: 0,896 Vlampunt: 42 °C (108 °F) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00067050 CAS nummer: 1569-02-4 EINECS: 216-374-5 UN: 1987 ADR: 3,III |