3-Methyl-2-cyclopenten-1-one ≥98% stabilised
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Stabilisatie:
Stabilised with hydroquinone (HQ) 0,10 %
A13993.09
A13993.18
A13993.09EA
80.8
EUR
A13993.09
A13993.18
3-Methyl-2-cyclopenten-1-one ≥98% stabilised
3-Methyl-2-cyclopenten-1-one
Stabilized with 0.1% hydroquinone
Purity: 98%
CAS No.: 2758-18-1
Molecular Formula: C6H8O
Formula Weight: 96.13
Storage Temperature: 2°C to 8°C
Flash Point: 65°(149°F)
Refractive Index: 1.4890
Odour: Pleasant
pH: 4.9
Melting Point: 2°
Boiling Point: 72-74°/15mm
Beilstein Reference: 1280476
MDL No.: MFCD00001403
Formule:
C₆H₈O Kookpunt: 72…74 °C (15 mmHg) Smeltpunt: 2 °C Dichtheid: 0,980 Vlampunt: 65 °C (149 °F) Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD00001403 CAS nummer: 2758-18-1 EINECS: 220-421-5 |