DL-Valine

Leverancier: BACHEM BIOCHEMICA
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: H-DL-Val-OH, (±)-Valine, 2-Amino-3-methylbutanoic acid

4031331.0025 4031331.0100
BACE4031331.0025EA 0 EUR
BACE4031331.0025 BACE4031331.0100
DL-Valine
DL-Valine
Formule: C₅H₁₁NO₂
Kookpunt: 217 °C (1013 hPa)
Smeltpunt: 86 °C
Dichtheid: 1,352 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004267
CAS nummer: 516-06-3
EINECS: 208-220-0
Merck Index: 12,10046

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR