5-Hydroxy-DL-tryptophan
Leverancier: Thermo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: DL-5-Hydroxytryptophan, 2-Amino-3-(5-hydroxy-1H-indol-3-yl)propanoic acid
AC12199.1 AC12199.10
MAYBAC12199.1EA 0 EUR
MAYBAC12199.1 MAYBAC12199.10
5-Hydroxy-DL-tryptophan
5-Hydroxy-DL-tryptophan
Formule: C₁₁H₁₂N₂O₃
Smeltpunt: 301 °C
Dichtheid: 1,405 g/cm³ (20 °C)
MDL Nummer: MFCD00005651
CAS nummer: 56-69-9
EINECS: 200-284-8
Merck Index: 13,04873