(±)-3-Butyne-2-ol 97%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
245310100 245310500
ACRO245310100EA 52.1 EUR
ACRO245310100 ACRO245310500
(±)-3-Butyne-2-ol 97%
(±)-3-Butyne-2-ol
Formule: C₄H₆O
Kookpunt: 104…108 °C (1013 hPa)
Smeltpunt: –12 °C
Dichtheid: 0,89 g/cm³
Vlampunt: 26 °C
MDL Nummer: MFCD00004541
CAS nummer: 2028-63-9
EINECS: 217-978-1
UN: 2929
ADR: 6.1,II
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4250 to 1.4270 (20°C, 589 nm)
GC ≥96.0 %