(±)-4-Phenyl-2-butanol ≥98%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
L05970.14 L05970.06
L05970.14EA 125 EUR
L05970.14 L05970.06
(±)-4-Phenyl-2-butanol ≥98%
(±)-4-Phenyl-2-butanol
Purity: 98% CAS No.: 2344-70-9 Molecular Formula: C10H14O Formula Weight: 150.22 Flash Point: 110°(230°F) Refractive Index: 1.5170 Boiling Point: 239° Beilstein Reference: 1908294 MDL No.: MFCD00044349
Formule: (C₆H₅)CH₂CH₂CH(OH)CH₃
MW: 150.22 g/mol
Kookpunt: 239 °C
Dichtheid: 0.980
Vlampunt: >110 °C (230 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00044349
CAS nummer: 2344-70-9
EINECS: 219-055-9