EDTA disodium salt standardised solution 0.0575 M (0.115N) in aqueous solution
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Idranal III, Edetic acid disodium salt, Kestranal® 2S, Ethylenedinitrilotetra-acetic acid disodium salt, Disodium dihydrogen ethylenediaminetetraacetate, Ethylenediaminetetra-acetic acid disodium salt
39782.K2 39782.K7
39782.K2EA 40.5 EUR
39782.K2 39782.K7
EDTA disodium salt standardised solution 0.0575 M (0.115N) in aqueous solution
EDTA disodium salt
CAS No.: 139-33-3 Molecular Formula: (NaO2CCH2)2NCH2CH2N(CH2CO2H)2 Formula Weight: 336.22 Physical Form: Liquid Odour: Odourless MDL No.: MFCD00070672
Formule: C₁₀H₁₄N₂Na₂O₈
MW: 336.21 g/mol
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00070672
CAS nummer: 139-33-3
EINECS: 205-358-3
Merck Index: 12,03556