Glycolic acid ∼66 - 70% in aqueous solution
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
2-Hydroxy acid, 2-Hydroxyacetic acid
L14980.AP
L14980.0F
L14980.APEA
24.8
EUR
L14980.AP
L14980.0F
Glycolic acid ∼66 - 70% in aqueous solution
Glycolic acid
CAS No.: 79-14-1
Molecular Formula: C2H4O3
Formula Weight: 76.05
Flash Point: None
Refractive Index: 1.411
Odour: Sweet
Boiling Point: 112°
Beilstein Reference: 1209322
Merck Reference: 14,4498
UN No.: 3265
MDL No.: MFCD00004312
Formule:
C₂H₄O₃ MW: 76.05 g/mol Kookpunt: 112 °C Dichtheid: 1.270 Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00004312 CAS nummer: 79-14-1 EINECS: 201-180-5 UN: 3265 ADR: 8 II Merck Index: 14,4498 |