Dimethylamine 40% (w/w) in aqueous solution
Leverancier: Thermo Fisher ScientificTotal Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
CAS No.: 124-40-3
Molecular Formula: (CH3)2NH
Formula Weight: 45.08
Flash Point: 15°(60°F)
Physical Form: Liquid in Ampoule
Density: 0.890
Refractive Index: 1.3700
Odour: Fish-like
Beilstein Reference: 605257
Merck Reference: 14,3228
UN No.: 1160
MDL No.: MFCD00008288
Formule:
C₂H₇N Dichtheid: 0,890 Vlampunt: 15 °C (60 °F) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00008288 CAS nummer: 124-40-3 EINECS: 204-697-4 UN: 1160 ADR: 3,II Merck Index: 14,3228 |