Hydrogeenbromide 48% w/w aqueous solution , 99,9999% Metals basis
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Hydrogen bromide
10991.AA 10991.AE 10991.AP
10991.AAEA 28.9 EUR
10991.AA 10991.AE 10991.AP
Hydrogeenbromide 48% w/w aqueous solution , 99,9999% Metals basis
Hydrogeenbromide
Purity: 99.9999% CAS No.: 10035-10-6 Molecular Formula: HBr Formula Weight: 80.92 Physical Form: Liquid Density: 1.490 Appearance: Colourless to faint yellow Beilstein Reference: 3587158 Merck Reference: 14,4778 UN No.: 1788 MDL No.: MFCD00011323
Formule: HBr
MW: 80.91 g/mol
Kookpunt: -66.7
Smeltpunt: -86
Dichtheid: 1.490
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011323
CAS nummer: 10035-10-6
EINECS: 233-113-0
UN: 1788
ADR: 8 II
Merck Index: 14,4778