Kaliumhydroxide 0.5 mol/l (0.5 N) in methanol, Titripur® volumetric solution, Supelco®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Potash caustic, Caustic potash
M-Clarity™ quality level = MQ100
1.09351.1000 1.09351.1003
1.09351.1000EA 70.2 EUR
1.09351.1000 1.09351.1003
Kaliumhydroxide 0.5 mol/l (0.5 N) in methanol, Titripur® volumetric solution, Supelco®
Kaliumhydroxide
Merck have introduced into the range of Titripur® Titrants some packs with additional technology - Supelco® SmartTitrants that use an RFID tag to provide a seamless data transfer between the smart titrator and the RFID tag on the label of the bottle of titrant. It will ensure that all information needed for method execution, result calculation and documentation is transferred to the titrator equipped with this technology. MettlerToledo Compact and Excellence titrators include this technology. Please see ordering table below for packs with RFID tag.
Formule: KOH
MW: 56.11 g/mol
Kookpunt: 1320
Smeltpunt: 360
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003553
CAS nummer: 1310-58-3
EINECS: 215-181-3
UN: 3286
ADR: 3 (6.1, 8) II
Merck Index: 12,07806
Order Now
Specification Test Results
Form liquid
Amount-of-substance concentration 0.4975 - 0.5025
Measurement uncertainty +/- 0.0015
Traceability NIST SRM