Natriumhydroxide 0.25 M NaOH (0.25N) volumetric solution, Fluka™
Leverancier: Honeywell Chemicals
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
31086-1L
31086-5L-VP
31086-1LEA
72.1
EUR
31086-1L
31086-5L-VP
Natriumhydroxide 0.25 M NaOH (0.25N) volumetric solution, Fluka™
Natriumhydroxide
Formule:
NaOH Kookpunt: 100 °C (1,013 hPa) Dichtheid: 1,000 g/cm³ (20 °C) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00003548 CAS nummer: 1310-73-2 EINECS: 215-185-5 UN: 1824 ADR: 8,III |
Specification Test Results
Factor | 0.999 - 1.001 |
BAM/EMPA reference material | effective |
BAM/EMPA Certificate Number | effective |
Traceable to NIST SRM | effective |