Bariumchloride standardised solution 0.1 N in aqueous solution
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
35582.K2 35582.K7
35582.K2EA 80.4 EUR
35582.K2 35582.K7
Bariumchloride standardised solution 0.1 N in aqueous solution
Bariumchloride
CAS No.: 10361-37-2 Molecular Formula: BaCl2 Formula Weight: 208.25 Physical Form: Liquid Appearance: Colourless Odour: Odourless MDL No.: MFCD00003445
Formule: BaCl₂
MW: 208.24 g/mol
Kookpunt: 1560
Smeltpunt: 963
Dichtheid: 1.034 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003445
CAS nummer: 10361-37-2
EINECS: 233-788-1
UN: 3287
ADR: 6.1 III
Merck Index: 12,00998