Tetrabutylammonium hydroxide 0.4 M in water for HPLC
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
420121000 420125000 420120025
ACRO420121000EA 78.3 EUR
ACRO420121000 ACRO420125000 ACRO420120025
Tetrabutylammonium hydroxide 0.4 M in water for HPLC
Tetrabutylammonium hydroxide
Formule: C₁₆H₃₇NO
MW: 259.48 g/mol
Smeltpunt: 29.85
Dichtheid: 0.968
Vlampunt: >110 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00009425
CAS nummer: 2052-49-5
EINECS: 218-147-6
UN: 3267
ADR: 8 II
Order Now
Specification Test Results
Appearance Clear solution
UV at 420 nm A: ≤0.15
Color scale ≤10 APHA
Titration with HCl 0.37 to 0.43 M
Bromide (Br) ≤500 ppm
Specificaties
Environmentally Preferable