Tetrabutylammonium hydroxide 1.0 M in aqueous solution for HPLC
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
43662.AE
43662.AC
43662.AP
43662.AEEA
199
EUR
43662.AE
43662.AC
43662.AP
Tetrabutylammonium hydroxide 1.0 M in aqueous solution for HPLC
Tetrabutylammonium hydroxide
CAS No.: 2052-49-5
Molecular Formula: C16H37NO
Formula Weight: 259.48
Flash Point: None
Physical Form: Liquid
Sensitivity: Air Sensitive
Beilstein Reference: 3571228
UN No.: 3267
MDL No.: MFCD00009425
Phase transfer catalyst, electrolyte in polarography, ion-pair extraction
Formule:
C₁₆H₃₇NO MW: 259.48 g/mol Smeltpunt: 29.85 Dichtheid: 0.968 Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00009425 CAS nummer: 2052-49-5 EINECS: 218-147-6 UN: 3267 ADR: 8 II |