Jood 0.5 mol/l (1 N) in aqueous solution, AVS TITRINORM volumetric solution
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
30914.238EA 31.1 EUR
30914.238 30914.295
Jood 0.5 mol/l (1 N) in aqueous solution, AVS TITRINORM volumetric solution
Jood
Formule: I₂
MW: 253.81 g/mol
Kookpunt: 184.3
Smeltpunt: 114
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011355
CAS nummer: 7553-56-2
EINECS: 231-442-4
UN: 3264
ADR: 8 II
Merck Index: 13,05036
Order Now
Specification Test Results
Titer (20°C; real value 0.2 % accuracy) 0.499 - 0.501 mol/l
NIST traceable Confirmed