Cerium(IV) sulphate 0.1 mol/l (0.1 N) in aqueous solution, Titripur® Reag. Ph. Eur. volumetric solution, Supelco®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Cerium disulphate, Ceric sulphate, Cerium(IV) sulphate
M-Clarity™ quality level = MQ100
1.09092.1000 1.09092.1003
1.09092.1000EA 92.91 EUR
1.09092.1000 1.09092.1003
Cerium(IV) sulphate 0.1 mol/l (0.1 N) in aqueous solution, Titripur® Reag. Ph. Eur. volumetric solution, Supelco®
Cerium(IV) sulphate
Merck have introduced into the range of Titripur® Titrants some packs with additional technology - Supelco® SmartTitrants that use an RFID tag to provide a seamless data transfer between the smart titrator and the RFID tag on the label of the bottle of titrant. It will ensure that all information needed for method execution, result calculation and documentation is transferred to the titrator equipped with this technology. MettlerToledo Compact and Excellence titrators include this technology. Please see ordering table below for packs with RFID tag.
Formule: Ce(SO₄)₂
MW: 332.24 g/mol
Smeltpunt: 195
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00148852
CAS nummer: 13590-82-4
EINECS: 237-029-5
UN: 3264
ADR: 8 III
Merck Index: 12,02036
Order Now
Specification Test Results
Form liquid
Amount-of-substance concentration 0.0995 - 0.1005
Measurement uncertainty +/- 0.0003
Traceability NIST SRM