Azido-PEG3-carboxytetramethylrhodamine 110 conjugate
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
J64510.X0
J64510.MB
J64510.X0EA
206
EUR
J64510.X0
J64510.MB
Azido-PEG3-carboxytetramethylrhodamine 110 conjugate
Azido-PEG3-carboxytetramethylrhodamine 110 conjugate
A water soluble azide functionalized- TAMRA dye. The water-soluble PEG spacer minimizes the interaction between the dye and the biomolecule to be labeled.
- Appearance: Dark red solid
- Solubility: Soluble in water
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...