Silver sulphate 10 g/l in sulphuric acid
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Disilver sulphate
279480010 279480025
ACRO279480010EA 116 EUR
ACRO279480010 ACRO279480025
Silver sulphate 10 g/l in sulphuric acid
Silver sulphate
Formule: Ag₂SO₄
MW: 311.8 g/mol
Kookpunt: 1085
Smeltpunt: 655
MDL Nummer: MFCD00003407
CAS nummer: 10294-26-5
EINECS: 233-653-7
UN: 1830
ADR: 8 II
Merck Index: 13,08603
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dd7afaa8e9ae24bf040d40aed0c93763ce897080&vid=ae7b50634b53507047974c2157e5e38c2eb90a20&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Clear colorless Solution
Solution contains 9.5 to 10.5 g/l Ag2SO4 in H2SO4 95-97%